Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATGAACTGAGATCAGACAGTTG[C/T]GCGGCATCTGGAGATAACATATTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM122B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM122B - family with sequence similarity 122B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166599.2 | 1654 | Silent Mutation | GCA,GCG | A,A 192 | NP_001160071.1 | |
NM_001166600.2 | 1654 | Silent Mutation | GCA,GCG | A,A 211 | NP_001160072.1 | |
NM_001170756.1 | 1654 | Silent Mutation | GCA,GCG | A,A 211 | NP_001164227.1 | |
NM_001170757.1 | 1654 | Silent Mutation | GCA,GCG | A,A 159 | NP_001164228.1 | |
NM_145284.5 | 1654 | Silent Mutation | GCA,GCG | A,A 193 | NP_660327.2 | |
XM_006724730.2 | 1654 | Silent Mutation | GCA,GCG | A,A 212 | XP_006724793.1 | |
XM_006724732.2 | 1654 | Silent Mutation | GCA,GCG | A,A 212 | XP_006724795.1 | |
XM_006724733.3 | 1654 | Silent Mutation | GCA,GCG | A,A 193 | XP_006724796.1 | |
XM_011531282.1 | 1654 | Silent Mutation | GCA,GCG | A,A 216 | XP_011529584.1 | |
XM_011531283.1 | 1654 | Silent Mutation | GCA,GCG | A,A 215 | XP_011529585.1 | |
XM_011531284.1 | 1654 | Silent Mutation | GCA,GCG | A,A 212 | XP_011529586.1 | |
XM_011531285.1 | 1654 | Silent Mutation | GCA,GCG | A,A 211 | XP_011529587.1 | |
XM_011531286.1 | 1654 | Silent Mutation | GCA,GCG | A,A 197 | XP_011529588.1 | |
XM_011531287.1 | 1654 | Silent Mutation | GCA,GCG | A,A 216 | XP_011529589.1 | |
XM_011531288.1 | 1654 | Silent Mutation | GCA,GCG | A,A 193 | XP_011529590.1 | |
XM_011531289.1 | 1654 | Silent Mutation | GCA,GCG | A,A 215 | XP_011529591.1 | |
XM_011531290.1 | 1654 | Silent Mutation | GCA,GCG | A,A 192 | XP_011529592.1 | |
XM_011531291.1 | 1654 | Silent Mutation | GCA,GCG | A,A 216 | XP_011529593.1 | |
XM_011531292.1 | 1654 | Silent Mutation | GCA,GCG | A,A 215 | XP_011529594.1 | |
XM_011531293.1 | 1654 | Silent Mutation | GCA,GCG | A,A 211 | XP_011529595.1 | |
XM_011531294.1 | 1654 | Silent Mutation | GCA,GCG | A,A 192 | XP_011529596.1 | |
XM_017029306.1 | 1654 | Silent Mutation | GCA,GCG | A,A 197 | XP_016884795.1 | |
XM_017029307.1 | 1654 | Silent Mutation | GCA,GCG | A,A 196 | XP_016884796.1 |