Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAACTGCTGATCAGTGACATTTGA[G/T]AAATTGCAAGAAAAATGCCATATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300170 MIM: 300202 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OFD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OFD1 - OFD1, centriole and centriolar satellite protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAPPC2 - trafficking protein particle complex 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011658.3 | Intron | NP_001011658.1 | ||||
NM_001128835.2 | Intron | NP_001122307.2 | ||||
NM_014563.5 | Intron | NP_055378.1 | ||||
XM_011545565.1 | Intron | XP_011543867.1 | ||||
XM_011545566.2 | Intron | XP_011543868.1 |