Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCACAACCCAGCTGTTTCTCACT[A/G]TCTAAACCTTTATGCCTGCTGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 309550 MIM: 300805 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FMR1 - fragile X mental retardation 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185075.1 | Intron | NP_001172004.1 | ||||
NM_001185076.1 | Intron | NP_001172005.1 | ||||
NM_001185081.1 | Intron | NP_001172010.1 | ||||
NM_001185082.1 | Intron | NP_001172011.1 | ||||
NM_002024.5 | Intron | NP_002015.1 |
FMR1-AS1 - FMR1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |