Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACGTCCAGGGCGCTGTTGCCTCC[C/G]GAGGCCCGGGGAAAGAGGAGGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300089 MIM: 300634 MIM: 300090 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IDH3G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IDH3G - isocitrate dehydrogenase 3 (NAD(+)) gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDZD4 - PDZ domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303512.1 | 1430 | Silent Mutation | TCC,TCG | S,S 387 | NP_001290441.1 | |
NM_001303513.1 | 1430 | Silent Mutation | TCC,TCG | S,S 285 | NP_001290442.1 | |
NM_001303514.1 | 1430 | Silent Mutation | TCC,TCG | S,S 272 | NP_001290443.1 | |
NM_001303515.1 | 1430 | Silent Mutation | TCC,TCG | S,S 306 | NP_001290444.1 | |
NM_001303516.1 | 1430 | Silent Mutation | TCC,TCG | S,S 300 | NP_001290445.1 | |
NM_032512.3 | 1430 | Silent Mutation | TCC,TCG | S,S 381 | NP_115901.2 |
SSR4 - signal sequence receptor subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |