Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTATACTGAGGTCTGAGTTCTGG[A/G]AGAAGCCTTTTCCACATTCACTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 314995 | ||||||||||||||||||||
Literature Links: |
ZNF41 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF41 - zinc finger protein 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324139.1 | 1945 | Missense Mutation | TCC,TTC | S,F 307 | NP_001311068.1 | |
NM_001324140.1 | 1945 | Missense Mutation | TCC,TTC | S,F 393 | NP_001311069.1 | |
NM_001324141.1 | 1945 | Missense Mutation | TCC,TTC | S,F 307 | NP_001311070.1 | |
NM_001324142.1 | 1945 | Missense Mutation | TCC,TTC | S,F 395 | NP_001311071.1 | |
NM_001324143.1 | 1945 | Missense Mutation | TCC,TTC | S,F 307 | NP_001311072.1 | |
NM_001324144.1 | 1945 | Missense Mutation | TCC,TTC | S,F 393 | NP_001311073.1 | |
NM_001324145.1 | 1945 | Missense Mutation | TCC,TTC | S,F 307 | NP_001311074.1 | |
NM_001324147.1 | 1945 | Missense Mutation | TCC,TTC | S,F 393 | NP_001311076.1 | |
NM_001324148.1 | 1945 | Missense Mutation | TCC,TTC | S,F 395 | NP_001311077.1 | |
NM_001324149.1 | 1945 | Missense Mutation | TCC,TTC | S,F 307 | NP_001311078.1 | |
NM_001324150.1 | 1945 | Missense Mutation | TCC,TTC | S,F 393 | NP_001311079.1 | |
NM_001324151.1 | 1945 | Missense Mutation | TCC,TTC | S,F 403 | NP_001311080.1 | |
NM_001324152.1 | 1945 | Missense Mutation | TCC,TTC | S,F 307 | NP_001311081.1 | |
NM_001324153.1 | 1945 | Missense Mutation | TCC,TTC | S,F 403 | NP_001311082.1 | |
NM_001324154.1 | 1945 | Missense Mutation | TCC,TTC | S,F 427 | NP_001311083.1 | |
NM_001324155.1 | 1945 | Missense Mutation | TCC,TTC | S,F 435 | NP_001311084.1 | |
NM_001324156.1 | 1945 | Missense Mutation | TCC,TTC | S,F 359 | NP_001311085.1 | |
NM_001324157.1 | 1945 | Missense Mutation | TCC,TTC | S,F 357 | NP_001311086.1 | |
NM_007130.3 | 1945 | Missense Mutation | TCC,TTC | S,F 393 | NP_009061.1 | |
NM_153380.3 | 1945 | Missense Mutation | TCC,TTC | S,F 393 | NP_700359.1 | |
XM_006724550.3 | 1945 | Missense Mutation | TCC,TTC | S,F 427 | XP_006724613.1 | |
XM_006724555.3 | 1945 | Missense Mutation | TCC,TTC | S,F 403 | XP_006724618.1 | |
XM_017029810.1 | 1945 | Missense Mutation | TCC,TTC | S,F 427 | XP_016885299.1 | |
XM_017029811.1 | 1945 | Missense Mutation | TCC,TTC | S,F 427 | XP_016885300.1 | |
XM_017029812.1 | 1945 | Missense Mutation | TCC,TTC | S,F 427 | XP_016885301.1 | |
XM_017029813.1 | 1945 | Missense Mutation | TCC,TTC | S,F 427 | XP_016885302.1 | |
XM_017029814.1 | 1945 | Missense Mutation | TCC,TTC | S,F 427 | XP_016885303.1 | |
XM_017029815.1 | 1945 | Missense Mutation | TCC,TTC | S,F 427 | XP_016885304.1 | |
XM_017029816.1 | 1945 | Missense Mutation | TCC,TTC | S,F 403 | XP_016885305.1 | |
XM_017029817.1 | 1945 | Missense Mutation | TCC,TTC | S,F 393 | XP_016885306.1 |