Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGATACCATTCCTGAACTCAGCC[A/G]ATTCACAGCATGCATTGATCTGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRG4 - adhesion G protein-coupled receptor G4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153834.3 | 190 | Missense Mutation | CAA,CGA | Q,R 54 | NP_722576.3 | |
XM_011531269.2 | 190 | Missense Mutation | CAA,CGA | Q,R 54 | XP_011529571.1 | |
XM_011531271.2 | 190 | UTR 5 | XP_011529573.1 |