Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGTTGTGGGCTCCAGGGTTCGA[A/G]CCACCGGCAACGTCTTGGGCTCCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300904 | ||||||||||||||||||||
Literature Links: |
IRS4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IRS4 - insulin receptor substrate 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003604.2 | 3792 | Silent Mutation | GGC,GGT | G,G 1184 | NP_003595.1 | |
XM_005262220.2 | 3792 | Silent Mutation | GGC,GGT | G,G 1184 | XP_005262277.1 | |
XM_006724713.3 | 3792 | Silent Mutation | GGC,GGT | G,G 1184 | XP_006724776.1 | |
XM_011531061.1 | 3792 | Silent Mutation | GGC,GGT | G,G 1184 | XP_011529363.1 |
LOC101928358 - uncharacterized LOC101928358 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |