Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCCAGAGACCACGGTGGCTATGA[C/T]TGGGGTGCGTGTGCCGGTGTGGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300443 | ||||||||||||||||||||
Literature Links: |
SLC7A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC7A3 - solute carrier family 7 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048164.2 | 1324 | Missense Mutation | ATC,GTC | I,V 384 | NP_001041629.1 | |
NM_032803.5 | 1324 | Missense Mutation | ATC,GTC | I,V 384 | NP_116192.4 | |
XM_017029912.1 | 1324 | Missense Mutation | ATC,GTC | I,V 384 | XP_016885401.1 |