Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGTCATCAAAACCACACTGATA[A/C]AAAGATTCCATTTTCCTCTTGCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300102 | ||||||||||||||||||||
Literature Links: |
PNPLA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PNPLA4 - patatin like phospholipase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142389.1 | 769 | Missense Mutation | TTG,TTT | L,F 234 | NP_001135861.1 | |
NM_001172672.1 | 769 | Missense Mutation | TTG,TTT | L,F 147 | NP_001166143.1 | |
NM_004650.2 | 769 | Missense Mutation | TTG,TTT | L,F 234 | NP_004641.1 | |
XM_011545588.1 | 769 | Missense Mutation | TTG,TTT | L,F 234 | XP_011543890.1 |