Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAATGCGCCACTGTTTCCATTTAC[C/G]TGGGAGCGAGACCGACCTTGATGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300741 | ||||||||||||||||||||
Literature Links: |
FAM120C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM120C - family with sequence similarity 120C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300788.1 | 2767 | Missense Mutation | ACG,AGG | T,R 895 | NP_001287717.1 | |
NM_017848.5 | 2767 | Missense Mutation | CAC,CAG | H,Q 1032 | NP_060318.4 | |
NM_198456.2 | 2767 | Intron | NP_940858.2 | |||
XM_006724589.3 | 2767 | Intron | XP_006724652.1 |