Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGCAGCTAGATGGAAAGTCCGCA[A/G]TGAAGTCAAACTCATTCTGCCCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300149 | ||||||||||||||||||||
Literature Links: |
CITED1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CITED1 - Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144885.1 | 1133 | Missense Mutation | ACT,ATT | T,I 212 | NP_001138357.1 | |
NM_001144886.1 | 1133 | Missense Mutation | ACT,ATT | T,I 186 | NP_001138358.1 | |
NM_001144887.1 | 1133 | Missense Mutation | ACT,ATT | T,I 186 | NP_001138359.1 | |
NM_004143.3 | 1133 | Missense Mutation | ACT,ATT | T,I 186 | NP_004134.2 | |
XM_011530958.1 | 1133 | Missense Mutation | ACT,ATT | T,I 212 | XP_011529260.1 |