Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCTTTACCTCCTGATTCTGGTG[A/G]CAAGAACCTTGTAATACAGAAATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300329 | ||||||||||||||||||||
Literature Links: |
NKAPP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NKAPP1 - NFKB activating protein pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM255A - transmembrane protein 255A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104544.1 | 758 | Intron | NP_001098014.1 | |||
NM_001104545.1 | 758 | Intron | NP_001098015.1 | |||
NM_017938.3 | 758 | Intron | NP_060408.3 | |||
XM_017029619.1 | 758 | Intron | XP_016885108.1 |
ZBTB33 - zinc finger and BTB domain containing 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184742.1 | 758 | Missense Mutation | GAC,GGC | D,G 144 | NP_001171671.1 | |
NM_006777.3 | 758 | Missense Mutation | GAC,GGC | D,G 144 | NP_006768.1 |