Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCGAAGACAAGAGTGATGATTC[A/G]GATGTTCCCGATGACAATAGTGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300369 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACRC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACRC - acidic repeat containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052957.4 | 1287 | Silent Mutation | TCA,TCG | S,S 322 | NP_443189.1 | |
XM_006724716.3 | 1287 | Silent Mutation | TCA,TCG | S,S 322 | XP_006724779.1 | |
XM_011531063.2 | 1287 | Silent Mutation | TCA,TCG | S,S 331 | XP_011529365.1 | |
XM_011531064.1 | 1287 | Silent Mutation | TCA,TCG | S,S 322 | XP_011529366.1 | |
XM_011531066.2 | 1287 | Silent Mutation | TCA,TCG | S,S 304 | XP_011529368.1 | |
XM_011531067.2 | 1287 | Silent Mutation | TCA,TCG | S,S 304 | XP_011529369.1 | |
XM_017029965.1 | 1287 | Intron | XP_016885454.1 |