Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCCTCTCCAGCCCTAGAAACCT[A/G]AGCTCTATAAATGTTGCCTCTCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300002 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARSD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARSD - arylsulfatase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001669.3 | 4485 | UTR 3 | NP_001660.2 | |||
NM_009589.3 | 4485 | Intron | NP_033667.2 | |||
XM_005274514.2 | 4485 | UTR 3 | XP_005274571.1 | |||
XM_005274515.2 | 4485 | UTR 3 | XP_005274572.1 |