Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GTTTCTCCAGAGTCGTTGGCCTCTC[A/T]GGCATGTGCCCTATGGCCCAAGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300694 MIM: 300353 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR223 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR223 - microRNA 223 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VSIG4 - V-set and immunoglobulin domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100431.1 | 1100 | Nonsense Mutation | AGA,TGA | R,* 231 | NP_001093901.1 | |
NM_001184830.1 | 1100 | UTR 3 | NP_001171759.1 | |||
NM_001184831.1 | 1100 | UTR 3 | NP_001171760.1 | |||
NM_001257403.1 | 1100 | Intron | NP_001244332.1 | |||
NM_007268.2 | 1100 | Nonsense Mutation | AGA,TGA | R,* 325 | NP_009199.1 | |
XM_017029251.1 | 1100 | Intron | XP_016884740.1 |