Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATGAGGTGATGGGAAATAGGGG[A/C]ATCCTCTTTTATAATCCAAGGCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300006 MIM: 300275 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CETN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CETN2 - centrin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NSDHL - NAD(P) dependent steroid dehydrogenase-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129765.1 | Intron | NP_001123237.1 | ||||
NM_015922.2 | Intron | NP_057006.1 | ||||
XM_011531178.2 | Intron | XP_011529480.1 | ||||
XM_017029564.1 | Intron | XP_016885053.1 |