Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTAGCATTATTCTACCCTTCTTT[C/T]CTTCCCTATGGTCTACTTTCAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 314990 | ||||||||||||||||||||
Literature Links: |
ZNF711 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF711 - zinc finger protein 711 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021998.4 | Intron | NP_068838.3 | ||||
XM_005262186.2 | Intron | XP_005262243.1 | ||||
XM_005262187.1 | Intron | XP_005262244.1 | ||||
XM_005262188.1 | Intron | XP_005262245.1 | ||||
XM_005262189.2 | Intron | XP_005262246.1 | ||||
XM_011531019.1 | Intron | XP_011529321.1 | ||||
XM_011531020.1 | Intron | XP_011529322.1 | ||||
XM_011531021.1 | Intron | XP_011529323.1 | ||||
XM_011531022.1 | Intron | XP_011529324.1 | ||||
XM_011531023.2 | Intron | XP_011529325.1 | ||||
XM_011531024.1 | Intron | XP_011529326.1 | ||||
XM_011531025.1 | Intron | XP_011529327.1 | ||||
XM_011531026.1 | Intron | XP_011529328.1 | ||||
XM_017029803.1 | Intron | XP_016885292.1 | ||||
XM_017029804.1 | Intron | XP_016885293.1 | ||||
XM_017029805.1 | Intron | XP_016885294.1 | ||||
XM_017029806.1 | Intron | XP_016885295.1 | ||||
XM_017029807.1 | Intron | XP_016885296.1 | ||||
XM_017029808.1 | Intron | XP_016885297.1 | ||||
XM_017029809.1 | Intron | XP_016885298.1 |