Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGATTGGCTTCAACTTTCTATTC[C/T]GAGGAGTACTAGGAGCCTGTCGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 311790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PFKFB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PFKFB1 - 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271804.1 | Intron | NP_001258733.1 | ||||
NM_001271805.1 | Intron | NP_001258734.1 | ||||
NM_002625.3 | Intron | NP_002616.2 | ||||
XM_017029576.1 | Intron | XP_016885065.1 | ||||
XM_017029577.1 | Intron | XP_016885066.1 | ||||
XM_017029578.1 | Intron | XP_016885067.1 |