Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATACTTCCACAGGTCCCACAACC[A/G]CAGCGAGGAGTTTCTCATTGCTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 313440 MIM: 305370 | ||||||||||||||||||||
Literature Links: |
MIR4769 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4769 - microRNA 4769 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYN1 - synapsin I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006950.3 | 451 | Intron | NP_008881.2 | |||
NM_133499.2 | 451 | Intron | NP_598006.1 |
TIMP1 - TIMP metallopeptidase inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003254.2 | 451 | Missense Mutation | CAC,CGC | H,R 102 | NP_003245.1 | |
XM_017029766.1 | 451 | Missense Mutation | ACA,GCA | T,A 26 | XP_016885255.1 |