Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGATAGTGTGAATATACTCTTC[C/T]TAGCAAGCTTTCATTCGATGTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300028 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZRSR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZRSR2 - zinc finger CCCH-type, RNA binding motif and serine/arginine rich 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005089.3 | Intron | NP_005080.1 | ||||
XM_005274597.2 | Intron | XP_005274654.2 | ||||
XM_011545589.2 | Intron | XP_011543891.2 | ||||
XM_011545590.2 | Intron | XP_011543892.2 | ||||
XM_017029881.1 | Intron | XP_016885370.1 | ||||
XM_017029882.1 | Intron | XP_016885371.1 | ||||
XM_017029883.1 | Intron | XP_016885372.1 |