Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATAAGAATTGGTATTCCTCATG[A/T]TGCTCTCCACCAATATGACAGGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300566 | ||||||||||||||||||||
Literature Links: |
LHFPL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LHFPL1 - lipoma HMGIC fusion partner-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178175.3 | 1151 | Missense Mutation | AAC,ATC | N,I 203 | NP_835469.1 | |
XM_011530943.2 | 1151 | Missense Mutation | AAC,ATC | N,I 226 | XP_011529245.1 | |
XM_011530944.2 | 1151 | Missense Mutation | AAC,ATC | N,I 203 | XP_011529246.1 | |
XM_011530946.2 | 1151 | Missense Mutation | AAC,ATC | N,I 170 | XP_011529248.1 | |
XM_017029485.1 | 1151 | Missense Mutation | AAC,ATC | N,I 226 | XP_016884974.1 |