Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCGAGAGGAGACCCAGGGTCTCA[A/G]GGTTGCTCACGCCACTGCAGCAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300097 MIM: 300153 | ||||||||||||||||||||
Literature Links: |
MAGEB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAGEB1 - MAGE family member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002363.4 | 240 | Missense Mutation | AAG,AGG | K,R 26 | NP_002354.2 | |
NM_177404.2 | 240 | Missense Mutation | AAG,AGG | K,R 26 | NP_796379.1 | |
NM_177415.2 | 240 | Missense Mutation | AAG,AGG | K,R 26 | NP_803134.1 |
MAGEB4 - MAGE family member B4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |