Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCATTGACTTGGCTCTCACTCAG[A/G]TAAGTTCCCATTTCACATTCACACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CXorf38 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CXorf38 - chromosome X open reading frame 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144970.2 | 746 | Silent Mutation | TAC,TAT | Y,Y 233 | NP_659407.1 | |
XM_005272589.2 | 746 | Silent Mutation | TAC,TAT | Y,Y 233 | XP_005272646.2 | |
XM_006724527.3 | 746 | Intron | XP_006724590.2 | |||
XM_006724528.2 | 746 | Silent Mutation | TAC,TAT | Y,Y 188 | XP_006724591.1 | |
XM_017029302.1 | 746 | Silent Mutation | TAC,TAT | Y,Y 244 | XP_016884791.1 | |
XM_017029303.1 | 746 | Silent Mutation | TAC,TAT | Y,Y 188 | XP_016884792.1 | |
XM_017029304.1 | 746 | Silent Mutation | TAC,TAT | Y,Y 114 | XP_016884793.1 | |
XM_017029305.1 | 746 | Silent Mutation | TAC,TAT | Y,Y 114 | XP_016884794.1 |