Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTGCTCCTCCTCAGGTCATAAA[C/G]GTGGAGCAGAGCTCTGTCTTCTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300332 MIM: 300084 | ||||||||||||||||||||
Literature Links: |
ITGB1BP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGB1BP2 - integrin subunit beta 1 binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303277.1 | 631 | Silent Mutation | AAC,AAG | N,K 152 | NP_001290206.1 | |
NM_012278.2 | 631 | Silent Mutation | AAC,AAG | N,K 275 | NP_036410.1 | |
XM_017029405.1 | 631 | Missense Mutation | AAC,AAG | N,K 174 | XP_016884894.1 |
NONO - non-POU domain containing, octamer-binding | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |