Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTCAGCACTTTCTCCACATGCT[A/G]TTTGAAATTAAGACATTTGGCAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300535 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OCRL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OCRL - OCRL, inositol polyphosphate-5-phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000276.3 | Intron | NP_000267.2 | ||||
NM_001318784.1 | Intron | NP_001305713.1 | ||||
NM_001587.3 | Intron | NP_001578.2 | ||||
XM_005262422.2 | Intron | XP_005262479.1 | ||||
XM_011531344.2 | Intron | XP_011529646.1 | ||||
XM_011531345.2 | Intron | XP_011529647.1 | ||||
XM_017029554.1 | Intron | XP_016885043.1 |