Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTTGAGTCCTCGTTCCCCTACT[C/T]GGGTGTCATATCCATGTGGCATTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300135 | ||||||||||||||||||||
Literature Links: |
ABCB7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCB7 - ATP binding cassette subfamily B member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271696.1 | 1753 | Missense Mutation | CAA,CGA | Q,R 601 | NP_001258625.1 | |
NM_001271697.1 | 1753 | Missense Mutation | CAA,CGA | Q,R 561 | NP_001258626.1 | |
NM_001271698.1 | 1753 | Missense Mutation | CAA,CGA | Q,R 575 | NP_001258627.1 | |
NM_001271699.1 | 1753 | Missense Mutation | CAA,CGA | Q,R 562 | NP_001258628.1 | |
NM_004299.4 | 1753 | Missense Mutation | CAA,CGA | Q,R 602 | NP_004290.2 |