Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAAATGGCAAAAATTACCTCCTT[C/T]GCAGGTGAGGTGCCACCCTTGACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300149 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CITED1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CITED1 - Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144885.1 | 630 | Silent Mutation | GCA,GCG | A,A 44 | NP_001138357.1 | |
NM_001144886.1 | 630 | Silent Mutation | GCA,GCG | A,A 18 | NP_001138358.1 | |
NM_001144887.1 | 630 | Silent Mutation | GCA,GCG | A,A 18 | NP_001138359.1 | |
NM_004143.3 | 630 | Silent Mutation | GCA,GCG | A,A 18 | NP_004134.2 | |
XM_011530958.1 | 630 | Silent Mutation | GCA,GCG | A,A 44 | XP_011529260.1 |