Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTAACAGGTTCTTGGGGAGGGAC[C/T]GATGTTTTCTGAGTTAGTTGAAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300182 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXorf38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXorf38 - chromosome X open reading frame 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MED14 - mediator complex subunit 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004229.3 | 4172 | Silent Mutation | TCA,TCG | S,S 1374 | NP_004220.2 | |
XM_005272701.2 | 4172 | Silent Mutation | TCA,TCG | S,S 1258 | XP_005272758.1 | |
XM_011544000.2 | 4172 | Silent Mutation | TCA,TCG | S,S 1258 | XP_011542302.1 | |
XM_017029962.1 | 4172 | Silent Mutation | TCA,TCG | S,S 1258 | XP_016885451.1 |