Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATTTCTATATTCCAGTCCTTACC[C/T]GCAGAGGCAAGCCGGCCTCTTCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300383 MIM: 311040 | ||||||||||||||||||||
Literature Links: |
CFP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFP - complement factor properdin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ELK1 - ELK1, ETS transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114123.2 | 869 | Missense Mutation | CAG,CGG | Q,R 218 | NP_001107595.1 | |
NM_001257168.1 | 869 | Intron | NP_001244097.1 | |||
NM_005229.4 | 869 | Missense Mutation | CAG,CGG | Q,R 218 | NP_005220.2 | |
XM_017029339.1 | 869 | Missense Mutation | CAG,CGG | Q,R 218 | XP_016884828.1 |