Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTACATATTAGGAACACATCGC[A/G]AAAATAGTTTAGCATCTTTCTTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300682 MIM: 300865 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00629 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00629 - long intergenic non-protein coding RNA 629 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR424 - microRNA 424 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR450A1 - microRNA 450a-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR450A2 - microRNA 450a-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR450B - microRNA 450b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR503 - microRNA 503 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR503HG - MIR503 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR542 - microRNA 542 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |