Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCATTGGCACTACCTCTCAACC[A/G]GTGCCGAATTCCCTGGGGAAAGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300463 MIM: 314375 MIM: 300249 | ||||||||||||||||||||
Literature Links: |
PQBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PQBP1 - polyglutamine binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC35A2 - solute carrier family 35 member A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIMM17B - translocase of inner mitochondrial membrane 17 homolog B (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167947.2 | 288 | Missense Mutation | CGG,TGG | R,W 97 | NP_001161419.1 | |
NM_005834.4 | 288 | Missense Mutation | CGG,TGG | R,W 47 | NP_005825.1 |