Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACATCCTCACTGGCTGTGCCAAA[A/G]GGACCATCCACTTCAATCCTGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300907 MIM: 300225 | ||||||||||||||||||||
Literature Links: |
CSTF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSTF2 - cleavage stimulation factor subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOX1 - NADPH oxidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271815.1 | 1358 | Silent Mutation | CCC,CCT | P,P 347 | NP_001258744.1 | |
NM_007052.4 | 1358 | Silent Mutation | CCC,CCT | P,P 384 | NP_008983.2 | |
NM_013955.2 | 1358 | Silent Mutation | CCC,CCT | P,P 384 | NP_039249.1 | |
XM_017029407.1 | 1358 | Silent Mutation | CCC,CCT | P,P 278 | XP_016884896.1 |