Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAGAGGAGGAGGAGGAAGAAGA[A/G]GAGGAGGAGGAAGAGGAGGAGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300922 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHIC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHIC1 - cysteine rich hydrophobic domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039840.2 | 420 | Silent Mutation | GAA,GAG | E,E 60 | NP_001034929.2 | |
NM_001300884.1 | 420 | Silent Mutation | GAA,GAG | E,E 60 | NP_001287813.1 | |
XM_017029582.1 | 420 | Silent Mutation | GAA,GAG | E,E 60 | XP_016885071.1 | |
XM_017029583.1 | 420 | Intron | XP_016885072.1 | |||
XM_017029584.1 | 420 | Silent Mutation | GAA,GAG | E,E 60 | XP_016885073.1 |
MAP2K4P1 - mitogen-activated protein kinase kinase 4 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |