Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAAGATAGCATGCCATAGACTAC[C/G]TTCTTTATGAAGCTATATCTATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300072 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP9X PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP9X - ubiquitin specific peptidase 9, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039590.2 | Intron | NP_001034679.2 | ||||
NM_001039591.2 | Intron | NP_001034680.2 | ||||
XM_005272675.4 | Intron | XP_005272732.1 | ||||
XM_005272676.4 | Intron | XP_005272733.1 |