Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGTGAAGTATTGAGTTTTATTT[C/T]AGGACCAAAGGACATAAACCAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM104B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM104B - family with sequence similarity 104 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166699.1 | 2669 | Missense Mutation | AAA,GAA | K,E 102 | NP_001160171.1 | |
NM_001166700.1 | 2669 | Intron | NP_001160172.1 | |||
NM_001166701.1 | 2669 | Intron | NP_001160173.1 | |||
NM_001166702.1 | 2669 | Intron | NP_001160174.1 | |||
NM_001166703.1 | 2669 | Intron | NP_001160175.1 | |||
NM_001166704.1 | 2669 | Intron | NP_001160176.1 | |||
NM_138362.3 | 2669 | Missense Mutation | AAA,GAA | K,E 101 | NP_612371.2 | |
XM_017029951.1 | 2669 | UTR 3 | XP_016885440.1 | |||
XM_017029952.1 | 2669 | UTR 3 | XP_016885441.1 | |||
XM_017029953.1 | 2669 | Missense Mutation | AAA,GAA | K,E 98 | XP_016885442.1 |