Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGACATAGTTAGCAGGACAGGGT[A/T]TGAGAGGGGTGTGAGGAGCAGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300303 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS6KA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPS6KA6 - ribosomal protein S6 kinase A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014496.4 | Intron | NP_055311.1 | ||||
XM_011530917.2 | Intron | XP_011529219.1 | ||||
XM_011530918.2 | Intron | XP_011529220.1 | ||||
XM_011530919.2 | Intron | XP_011529221.1 | ||||
XM_011530920.2 | Intron | XP_011529222.1 | ||||
XM_017029423.1 | Intron | XP_016884912.1 | ||||
XM_017029424.1 | Intron | XP_016884913.1 | ||||
XM_017029425.1 | Intron | XP_016884914.1 | ||||
XM_017029426.1 | Intron | XP_016884915.1 |