Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTTTGGGTGAGGAAGGGGGTGA[A/G]GTTTTGAGTGGAAGCCATGAGAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300948 | ||||||||||||||||||||
Literature Links: |
PCYT1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCYT1B - phosphate cytidylyltransferase 1, choline, beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163264.1 | 1093 | Silent Mutation | ACC,ACT | T,T 327 | NP_001156736.1 | |
NM_001163265.1 | 1093 | Intron | NP_001156737.1 | |||
NM_004845.4 | 1093 | Silent Mutation | ACC,ACT | T,T 345 | NP_004836.2 | |
XM_017029977.1 | 1093 | Silent Mutation | ACC,ACT | T,T 249 | XP_016885466.1 |