Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTTTAAACTTGGCTTAATAAGA[C/G]AATTTCCTACTGCTTCCTTTGTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300358 | ||||||||||||||||||||
Literature Links: |
WNK3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WNK3 - WNK lysine deficient protein kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002838.3 | 5433 | Missense Mutation | TCT,TGT | S,C 1608 | NP_001002838.1 | |
NM_020922.4 | 5433 | Missense Mutation | TCT,TGT | S,C 1665 | NP_065973.2 | |
XM_006724593.3 | 5433 | Missense Mutation | TCT,TGT | S,C 1608 | XP_006724656.1 | |
XM_011530802.2 | 5433 | Missense Mutation | TCT,TGT | S,C 1618 | XP_011529104.1 | |
XM_017029741.1 | 5433 | Missense Mutation | TCT,TGT | S,C 1665 | XP_016885230.1 | |
XM_017029742.1 | 5433 | Missense Mutation | TCT,TGT | S,C 1665 | XP_016885231.1 | |
XM_017029743.1 | 5433 | Missense Mutation | TCT,TGT | S,C 1665 | XP_016885232.1 | |
XM_017029744.1 | 5433 | Missense Mutation | TCT,TGT | S,C 1665 | XP_016885233.1 | |
XM_017029745.1 | 5433 | Missense Mutation | TCT,TGT | S,C 1655 | XP_016885234.1 |