Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACTGCACGATGCTGTCCCGGAG[C/T]TCCCAGAAGCGGGAGTCCAATTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300708 | ||||||||||||||||||||
Literature Links: |
FAM58A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM58A - family with sequence similarity 58 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130997.2 | 976 | Intron | NP_001124469.1 | |||
NM_152274.4 | 976 | Intron | NP_689487.2 | |||
XM_005277920.4 | 976 | Intron | XP_005277977.1 | |||
XM_005277921.4 | 976 | Intron | XP_005277978.1 | |||
XM_011531214.2 | 976 | Silent Mutation | GAA,GAG | E,E 74 | XP_011529516.1 | |
XM_011531215.2 | 976 | Silent Mutation | GAA,GAG | E,E 74 | XP_011529517.1 |
LOC105373383 - uncharacterized LOC105373383 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |