Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGGTTTGGCAAGAGCTAAAAC[A/G]GTGCCTGGGGAAGGGGGCTTCATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300713 MIM: 300295 | ||||||||||||||||||||
Literature Links: |
OTUD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OTUD5 - OTU deubiquitinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136157.1 | 2251 | Silent Mutation | ACC,ACT | T,T 475 | NP_001129629.1 | |
NM_001136158.1 | 2251 | Silent Mutation | ACC,ACT | T,T 475 | NP_001129630.1 | |
NM_001136159.1 | 2251 | Silent Mutation | ACC,ACT | T,T 258 | NP_001129631.1 | |
NM_017602.3 | 2251 | Silent Mutation | ACC,ACT | T,T 480 | NP_060072.1 | |
XM_005272622.3 | 2251 | Silent Mutation | ACC,ACT | T,T 475 | XP_005272679.1 | |
XM_005278039.3 | 2251 | Silent Mutation | ACC,ACT | T,T 480 | XP_005278096.1 | |
XM_005278042.4 | 2251 | Silent Mutation | ACC,ACT | T,T 263 | XP_005278099.1 | |
XM_006724537.2 | 2251 | Silent Mutation | ACC,ACT | T,T 480 | XP_006724600.1 | |
XM_011543932.1 | 2251 | Silent Mutation | ACC,ACT | T,T 480 | XP_011542234.1 | |
XM_011543934.2 | 2251 | Silent Mutation | ACC,ACT | T,T 242 | XP_011542236.1 | |
XM_017029628.1 | 2251 | Silent Mutation | ACC,ACT | T,T 475 | XP_016885117.1 | |
XM_017029629.1 | 2251 | Silent Mutation | ACC,ACT | T,T 263 | XP_016885118.1 | |
XM_017029630.1 | 2251 | Silent Mutation | ACC,ACT | T,T 263 | XP_016885119.1 |
PIM2 - Pim-2 proto-oncogene, serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |