Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCTTAGGTTCGTGGGGTCCACC[A/G]TGGCAGGAGGCTGAGCAGGGGGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300383 MIM: 311040 MIM: 313440 | ||||||||||||||||||||
Literature Links: |
CFP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFP - complement factor properdin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145252.1 | 756 | Silent Mutation | CAC,CAT | H,H 210 | NP_001138724.1 | |
NM_002621.2 | 756 | Silent Mutation | CAC,CAT | H,H 210 | NP_002612.1 | |
XM_017029575.1 | 756 | Silent Mutation | CAC,CAT | H,H 75 | XP_016885064.1 |
ELK1 - ELK1, ETS transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYN1 - synapsin I | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |