Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTACCAGTTGGGAATTTTGGGAC[C/T]GTGAGTGTGGGAGAATCACTAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300224 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGED1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAGED1 - MAGE family member D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005332.1 | Intron | NP_001005332.1 | ||||
NM_001005333.1 | Intron | NP_001005333.1 | ||||
NM_006986.3 | Intron | NP_008917.3 | ||||
XM_011530835.2 | Intron | XP_011529137.1 | ||||
XM_017029978.1 | Intron | XP_016885467.1 | ||||
XM_017029979.1 | Intron | XP_016885468.1 | ||||
XM_017029980.1 | Intron | XP_016885469.1 |