Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTTGTGAACATCTCTGGCAACA[A/G]TGAGTCTACCTTGAACTGTTCACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300034 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGTR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGTR2 - angiotensin II receptor type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000686.4 | 191 | Missense Mutation | AAT,AGT | N,S 29 | NP_000677.2 | |
XM_011537533.1 | 191 | Missense Mutation | AAT,AGT | N,S 29 | XP_011535835.1 |