Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATTTCGCATCCTACACCAGACA[A/C]CAGATTCTCAAGTTTACTTGTCAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TBC1D8B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBC1D8B - TBC1 domain family member 8B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017752.2 | 373 | Missense Mutation | ACA,CCA | T,P 70 | NP_060222.2 | |
NM_198881.1 | 373 | Missense Mutation | ACA,CCA | T,P 70 | NP_942582.1 | |
XM_006724669.1 | 373 | Missense Mutation | ACA,CCA | T,P 70 | XP_006724732.1 | |
XM_006724670.3 | 373 | UTR 5 | XP_006724733.1 |