Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGATGGTCATGGTGAGGGGGATC[A/T]GTGCTGACTGCCCGCAAACTATGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300691 | ||||||||||||||||||||
Literature Links: |
BEX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BEX2 - brain expressed X-linked 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168399.1 | 535 | Silent Mutation | ACA,ACT | T,T 145 | NP_001161871.1 | |
NM_001168400.1 | 535 | Silent Mutation | ACA,ACT | T,T 144 | NP_001161872.1 | |
NM_001168401.1 | 535 | Silent Mutation | ACA,ACT | T,T 113 | NP_001161873.1 | |
NM_032621.3 | 535 | Silent Mutation | ACA,ACT | T,T 113 | NP_116010.1 |