Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCATGACGGAGCTGCTGCCACC[A/C]CAGCAGATCTGCTCGCCTTGCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300540 MIM: 300370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAUS7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAUS7 - HAUS augmin like complex subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017518.7 | 1091 | Missense Mutation | TGG,TGT | W,C 349 | NP_059988.3 |
TREX2 - three prime repair exonuclease 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |