Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAATTTTACCATTGGAGTTTTA[C/G]TTGCCCGTTTCTTCTTTTCTGCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300012 | ||||||||||||||||||||
Literature Links: |
SMARCA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SMARCA1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282874.1 | 3263 | Missense Mutation | ACT,AGT | T,S 1043 | NP_001269803.1 | |
NM_001282875.1 | 3263 | Missense Mutation | ACT,AGT | T,S 1031 | NP_001269804.1 | |
NM_003069.4 | 3263 | Missense Mutation | ACT,AGT | T,S 1043 | NP_003060.2 | |
XM_005262461.2 | 3263 | Missense Mutation | ACT,AGT | T,S 1043 | XP_005262518.1 | |
XM_005262462.2 | 3263 | Missense Mutation | ACT,AGT | T,S 1043 | XP_005262519.1 | |
XM_006724782.2 | 3263 | Missense Mutation | ACT,AGT | T,S 1031 | XP_006724845.1 | |
XM_017029750.1 | 3263 | Missense Mutation | ACT,AGT | T,S 1031 | XP_016885239.1 | |
XM_017029751.1 | 3263 | Missense Mutation | ACT,AGT | T,S 1031 | XP_016885240.1 |