Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGGCAGCTGGATCTTGCTGGG[A/G]TCGAAGCAGTTGGATTCCATGATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300403 MIM: 300080 | ||||||||||||||||||||
Literature Links: |
NDUFB11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFB11 - NADH:ubiquinone oxidoreductase subunit B11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135998.2 | 960 | Silent Mutation | GAC,GAT | D,D 143 | NP_001129470.1 | |
NM_019056.6 | 960 | Silent Mutation | GAC,GAT | D,D 153 | NP_061929.2 |
RBM10 - RNA binding motif protein 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |