Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGTGAACTCATCTGAAACCAAT[A/G]CAAGCATTTTTACAGTGTGGCTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300562 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLITRK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLITRK4 - SLIT and NTRK like family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184749.2 | 4208 | UTR 3 | NP_001171678.1 | |||
NM_001184750.2 | 4208 | UTR 3 | NP_001171679.1 | |||
NM_173078.4 | 4208 | UTR 3 | NP_775101.1 | |||
XM_005262365.4 | 4208 | Intron | XP_005262422.1 | |||
XM_011531265.2 | 4208 | Intron | XP_011529567.1 |