Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAAGTGTGCGACAAGTCCTACAC[G/T]CACCCGAGCTCCCTGCGCAAACACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300265 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZIC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZIC3 - Zic family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003413.3 | 1699 | Silent Mutation | ACG,ACT | T,T 398 | NP_003404.1 | |
XM_017029802.1 | 1699 | Silent Mutation | ACG,ACT | T,T 398 | XP_016885291.1 |